ZM-323881 ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

BMLEI3850001
ENZOBMLEI3850001EA 1025 NOK
ENZOBMLEI3850001
ZM-323881 ≥98% (by TLC)
ZM-323881

VEGFR inhibitor


Synthetic.

Potent and selective inhibitor of the kinase activity of the human vascular endothelial growth factor receptor 2 (VEGFR2/KDR). Selectively inhibits VEGFR2 over VEGFR1 and other receptor tyrosine kinases with an IC50 = 2nM. Useful tool for exploring the biology of VEGFR receptors.

MW: 375.4 g/mol
CAS-nummer: 193001-14-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1abfd7937b0a8f6acfba2e1090c2f6093047ea55&vid=ff8289818e5e7ac0e451cc971254441da20111c6&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR