cis-4,7,10,13,16,19-Docosahexaenoic acid ≥98%, colourless oil, Ultrapure

Leverandør: ENZO LIFE SCIENCES

BMLFA0020100 BMLFA0021000
ENZOBMLFA0020100EA 1025 NOK
ENZOBMLFA0020100 ENZOBMLFA0021000
cis-4,7,10,13,16,19-Docosahexaenoic acid ≥98%, colourless oil, Ultrapure
cis-4,7,10,13,16,19-Docosahexaenoic acid

Polyunsaturated fatty acid.


Soluble in ethanol, hexane and methylene chloride.

Formel: C₂₂H₃₂O₂
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00065722
CAS-nummer: 6217-54-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405d38db02874f4083895c0237a0f9afd14a3b10&vid=44c7fe7b7b208769f34a5de99a18dcb81478fae4&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98%

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR