Anti-FN1 Mouse Monoclonal Antibody
Leverandør: ENZO LIFE SCIENCES
BMLFG60200100
ENZOBMLFG60200100EA 2550 NOK
ENZOBMLFG60200100
Anti-FN1 Mouse Monoclonal Antibody
Antistoffer
Fibronectins are high molecular weight, disulphide-linked, dimeric cell adhesion glycoproteins found in basement membranes and in the interstitial connective tissue matrix. A single fibronectin gene is subject to alternative splicing in a cell-type-, development- and age-regulated manner which gives rise to multiple molecular forms. In addition to their prominent role in adhesion, fibronectins have been reported to mediate various aspects of cellular interaction, including migration during development and wound-healing, haemostasis, and the regulation of cell growth and differentiation.More Product Information
New Product