3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: MCI-186, 1-Phenyl-3-methyl-5-pyrazolone, 3-Methyl-1-phenyl-1H-pyrazol-5(4H)-one, Edaravone

BMLFR1030050 BMLFR1030250
ENZOBMLFR1030050EA 790 NOK
ENZOBMLFR1030050 ENZOBMLFR1030250
3-Methyl-1-phenyl-2-pyrazolin-5-one ≥98% (by HPLC)
3-Methyl-1-phenyl-2-pyrazolin-5-one

A free radical scavenger and antioxidant. Displays beneficial effects in a rat model of ischemic brain damage.


Soluble in DMSO.

Formel: C₁₀H₁₀N₂O
MDL nummer: MFCD00003138
CAS-nummer: 89-25-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6f0ac378de9f4d1748ec0c286568f5c7d13be14&vid=63fb9373f5df4649427deb61fe6e74e8b589cc57&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)
Appearance White to beige to yellow powder

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR