Resveratrol 98 TLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

Synonymer: trans-3,4,5-Trihydroxystilbene, (E)-5-(4-Hydroxystyryl)benzene-1,3-diol

BMLFR1040100
ENZOBMLFR1040100EA 790 NOK
ENZOBMLFR1040100
Resveratrol 98 TLC_ASSAY_METHOD
Resveratrol

Antioxidant. SIRT1 activator.


Phenolic antioxidant found in grapes and wine. Potent activator of human deacetylase SIRT1 (maximum effect at 100-200μM). Increases longevity in yeast. Displays antiproliferative effect on human breast epithelial cells. Displays a variety of pharmacological activities including: estrogen receptor agonist activity, inhibition of ribonucleotide reductase and DNA synthesis, inhibition of eicosanoid synthesis, cell cycle arrest at S/G2 phase and induction of apoptosis. Inhibits PKD in vitro (IC50=200μM) and in vivo (IC50=800μM), but not PKC isoforms. Protects against 4-hydroxynonenal (4-HNE) induced oxidative stress and apoptosis. Shows cancer chemopreventive activity. Specific inhibitor of cyclooxygenase-1 (COX-1). Inhibits the hydroperoxidase activity of COX-1. 

Formel: C₁₄H₁₂O₃
MW: 228.25 g/mol
Smeltepunkt: 260
MDL nummer: MFCD00133799
CAS-nummer: 501-36-0
Merck Indeks: 12,08326
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=391f2c3c9be94b56f8f5ec16e9947769561f9735&vid=c523d3af4258764798e3b170923b095a446a333c&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR