Bordetella pertussis toxin
Leverandør: ENZO LIFE SCIENCES
BMLG1010050
ENZOBMLG1010050EA 5050 NOK
ENZOBMLG1010050
Bordetella pertussis toxin
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Seren
Bordetella pertussis toxin (Islet-activating protein, PTX) from Bordetella pertussis, strain NCTC 10739. is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies, LINE assay.
  • High purity, native protein available as >98% pure
  • Minimal lot-to-lot variation ensures reproducible results
  • Lyophilised format for extended shelf-life