Clostridium difficile toxin A
Leverandør: ENZO LIFE SCIENCES
BMLG1400050
ENZOBMLG1400050EA 5600 NOK
ENZOBMLG1400050
Clostridium difficile toxin A
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Seren
Clostridium difficile toxin A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
  • Minimal lot to lot variation
  • Single lot bulk quantities available
  • Lyophilised for ease of handling and long term stability