Clostridium difficile toxin B
Leverandør: ENZO LIFE SCIENCES
BMLG1500050
ENZOBMLG1500050EA 13925 NOK
ENZOBMLG1500050
Clostridium difficile toxin B
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Seren
Clostridium difficile toxin B from Clostridium difficile, strain VPI10463 is a high purity toxin applicable to ELISA, antibody capture, cytotoxicity studies and LINE assay.
  • High potency retained following inactivation
  • Minimal lot-to-lot variation
  • Lyophilised for ease of handling and long term stability