Clostridium difficile toxoid B
Leverandør: ENZO LIFE SCIENCES
BMLG1550050
ENZOBMLG1550050EA 13475 NOK
ENZOBMLG1550050
Clostridium difficile toxoid B
Mikrobiologiske testsystemer Antigen-antistoff tester Ag-AK, Seren
Clostridium difficile toxoid A from Clostridium difficile, strain VPI10463 is a high potency toxin applicable to ELISA, antibody capture, cytotoxicity studies, immunogen and LINE assay.
  • Minimal lot-to-lot variation
  • Inactivated by formaldehyde
  • Lyophilised for ease of handling and long term stability