13-cis-Retinoic acid ≥95% (by TLC)

Leverandør: ENZO LIFE SCIENCES
Danger

Synonymer: Isotretinoin, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethyl-1-cyclohexen-1-yl)-2,4,6,8-nonatetraenoic Acid, (2Z,4E,6E,8E)-3,7-Dimethyl-9-(2,6,6-trimethylcyclohex-1-en-1-yl)nona-2,4,6,8-tetraenoic acid

BMLGR1020500
ENZOBMLGR1020500EA 2550 NOK
ENZOBMLGR1020500
13-cis-Retinoic acid ≥95% (by TLC)
13-cis-Retinoic acid

Retinoic acid receptor ligand.


Retinoic acid is the active metabolite of vitamin A. It is a potent modulator of growth and differentiation exerting its effects by binding to nuclear retinoic acid receptors (RARs) which directly regulate gene expression. All trans retinoic acid binds to RAR α, β and γ subtypes with Kd values of 0.2, 0.4 and 0.2 nM respectively. RARs function as ligand-dependent transcription factors. Retinoic Acid also promotes the differentiation of mouse embryonic stem cells into adipocytes, neurons and glia in vitro. It guides development of the posterior part of the embryo.


Soluble in DMSO (25 mg/ml) or ethanol (10 mg/ml).

Formel: C₂₀H₂₈O₂
MW: 300,44 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00079542
CAS-nummer: 4759-48-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f059265fe4a41ba59296f9e936e4bb832a28942d&vid=2f5f9871045d984db02c76a15ec32817c52de7b0&prid=web&clid=SAPDAM
New Product

Bestill nå

Spesifikasjon Testresultater

Purity ≥95%
Appearance Orange solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR