GW-9662 98 HPLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: GW9662, 2-Chloro-5-nitro-N-phenylbenzamide

BMLGR2340010 BMLGR2340050
ENZOBMLGR2340010EA 1375 NOK
ENZOBMLGR2340010 ENZOBMLGR2340050
GW-9662 98 HPLC_ASSAY_METHOD
GW-9662

PPAR ligand.


Potent, selective and irreversible PPARγ antagonist. Blocks the PPARγ-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 µM. Can be used to antagonise the effects of PPARγ agonists in a variety of cell types.


Soluble in DMSO (50 mg/ml), 100% ethanol (10 mg/ml) or dimethyl formamide. Insoluble in water.

Formel: C₁₃H₉N₂O₃Cl
MW: 276.7 g/mol
Smeltepunkt: 155…157 °C
MDL nummer: MFCD01215270
CAS-nummer: 22978-25-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2e921714d1c9b480af8862b60e9530751471116d&vid=930ede030b5767df965756e8f6c6b3f9981d185b&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR