Z-Guggulsterone ≥98%

Leverandør: ENZO LIFE SCIENCES

BMLGR2350005
ENZOBMLGR2350005EA 1975 NOK
ENZOBMLGR2350005
Z-Guggulsterone ≥98%
Z-Guggulsterone

A guggul tree steroid well known for its lipid lowering properties. Acts as a selective antagonist at farnesoid X receptors (FXRs). Inhibits FXR transactivation (IC50=10µM).


  • FXR inhibitor


White solid.


Soluble in DMSO.

Formel: C₂₁H₂₈O₂
Lagringstemperatur: Fryseskap
CAS-nummer: 39025-23-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b316ae0fb828fa3cc20957b2a385241fa0e02996&vid=3ff4e85a33d8a11101c0ee0a5c6fda14e2ae51d7&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR