β-Lapachone ≥98%

Leverandør: ENZO LIFE SCIENCES

BMLGR3080005 BMLGR3080025
ENZOBMLGR3080005EA 1050 NOK
ENZOBMLGR3080005 ENZOBMLGR3080025
β-Lapachone ≥98%
beta-Lapachone

Topoisomerase inhibitor


Novel DNA topoisomerase I inhibitor. Unlike camptothecin, beta-lapachone does not stabilize the cleavable complex, indicating a novel mode of action. Induces apoptosis in HL-60 and human prostate cancer cells via a p53-independent mechanism and blocks the cell cycle at G0/G1.

Formel: C₁₅H₁₄O₃
MDL nummer: MFCD01712233
CAS-nummer: 4707-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=137fe9429303180eead3f422ecfeb1db9486cd07&vid=9c4fdd99ef57a4ab26890c4abc4ffaf8e875a1ad&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR