Pifithrin-α hydrobromide 98 TLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

Synonymer: Pifithrin-α HBr, Pifithrin-alpha, 2-(2-Imino-4,5,6,7-tetrahydrobenzothiazol-3-yl)-1-p-tolylethanone Hydrobromide

BMLGR3250005 BMLGR3250025
ENZOBMLGR3250005EA 1025 NOK
ENZOBMLGR3250005 ENZOBMLGR3250025
Pifithrin-α hydrobromide 98 TLC_ASSAY_METHOD
Pifithrin-alpha hydrobromide

Chemical inhibitor of p53 that protects mice from the side effects of cancer therapy. Reversibly blocks p53-dependent transcriptional activation and apoptosis. Protects against neuronal death in models of stroke and neurodegenerative disorders. Suppresses self-renewal of embryonic stem cells.


  • p53 inhibitor


White solid.


Soluble in DMSO (>25mg/ml) or methanol.

Formel: C₁₆H₁₈N₂OS·HBr
MW: 286.4·80.9 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00417851
CAS-nummer: 63208-82-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d270677fb1594cb6d3f46cbfb522b947cc3ab15e&vid=4237ff6b9af9bb33d19589687d711179724e9933&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR