Cathepsin S Assay Buffer
Leverandør: ENZO LIFE SCIENCES
BMLKI4310020
ENZOBMLKI4310020EA 620 NOK
ENZOBMLKI4310020
Cathepsin S Assay Buffer
Buffere Buffere, assays
50mM potassium phosphate buffer, pH 6,5, 50 mM NaCl, 2 mM EDTA, 0,01% Triton X-100®, 0,5 mM DTT.