WEB-2086 98 HPLC_ASSAY_METHOD

Leverandør: ENZO LIFE SCIENCES

BMLL1370005
ENZOBMLL1370005EA 5300 NOK
ENZOBMLL1370005
WEB-2086 98 HPLC_ASSAY_METHOD
WEB-2086

PAF receptor antagonist. Inhibits PAF-induced human platelet and neutrophil aggregation in vitro (IC50=0.17 and 0.36µM respectively). In vivo it dose dependently blocks eosinophil activation and displays antiallergic and anti-inflammatory activity in various models. Induces differentiation and triggers apoptosis and in a variety of cell lines.


  • PAF receptor ligand


Beige solid.


Soluble in DMSO or ethanol.

Formel: C₂₂H₂₂ClN₅O₂S
MW: 456 g/mol
Lagringstemperatur: Kjøleskap
CAS-nummer: 105219-56-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d1b608b9ff02db91d952a3d1d4aff4206ba91857&vid=e835495639e110248c0cfb0d978a44c960dab719&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR