2-Amino-6-(trifluoromethoxy)benzothiazole ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: 6-(Trifluoromethoxy)benzo[d]thiazol-2-amine, Riluzole

BMLNA1380020
ENZOBMLNA1380020EA 995 NOK
ENZOBMLNA1380020
2-Amino-6-(trifluoromethoxy)benzothiazole ≥98% (by TLC)
2-Amino-6-(trifluoromethoxy)benzothiazole

Na+ channel blocker, K+ channel activator


Glutamate release inhibitor. Neuroprotective. Blocks the inactivated state of voltage-gated neuronal Na+ channels at very low concentrations (about 0.1µM) and activates the TREK family of two-pore domain potassium channels.

Formel: C₈H₅F₃N₂OS
MW: 234.20 g/mol
MDL nummer: MFCD00210213
CAS-nummer: 1744-22-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8204234abdd45b1be789a6b76603cef17365ab46&vid=7e537d94fd541b13729a7bc3f9d5db5e9f0c9b06&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR