Citalopram hydrobromide ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: 1-[3-(Dimethylamino)propyl]-1-(4-fluorophenyl)-1,3-dihydroisobenzofuran-5-carbonitrile Hydrobromide

BMLNS1120050
ENZOBMLNS1120050EA 5300 NOK
ENZOBMLNS1120050
Citalopram hydrobromide ≥98% (by TLC)
Citalopram hydrobromide

Potent and selective serotonin uptake inhibitor (Ki=5.4nM). Modifies gene expression profile in Alzheimer lymphocytes. Clinically useful antidepressant.


  • Serotonin transporter inhibitor


White solid.


Soluble in DMSO (25mg/ml), ethanol (20mg/ml) or water (4mg/ml).

Formel: C₂₀H₂₁FN₂O
MW: 405.31 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD02101306
CAS-nummer: 59729-32-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=101bb6b53a45f9f5742d9406abd506318dde18d0&vid=bde4ad768708cef2f6cddf4766d6fccb737f141e&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR