3-Tropanyl-3,5-dichlorobenzoate ≥99% (by HPLC), white solid

Leverandør: ENZO LIFE SCIENCES
Danger

Synonymer: MDL-72222, Mdl 72222, rel-(1R,5R)-8-Methyl-8-azabicyclo[3.2.1]octan-3-yl 3,5-dichlorobenzoate, Bemesetron

BMLNS5300050
ENZOBMLNS5300050EA 5300 NOK
ENZOBMLNS5300050
3-Tropanyl-3,5-dichlorobenzoate ≥99% (by HPLC), white solid
3-Tropanyl-3,5-dichlorobenzoate

Serotonin receptor ligand


  • Serotonin 5-HT3 antagonist.


Soluble in DMSO.

Formel: C₁₅H₁₇Cl₂NO₂
MW: 314.206 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00865533
CAS-nummer: 40796-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2bace0fea31dab618d8589eefe4220f1155bde59&vid=d9646f8b9c8d24bebd66255a697c07429187b1fe&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR