Z-AAD-CMK
Leverandør: ENZO LIFE SCIENCES
BMLP1650001
ENZOBMLP1650001EA 2025 NOK
ENZOBMLP1650001
Z-AAD-CMK
Proteiner
Caspase inhibitor
Irreversible, cell-permeable inhibitor of granzyme B (ID50=0,3 µM). Does not inhibit granzyme A. Inhibition of granzyme B with Z-AAD-CMK prevented caspase-3 activation in T cells.