Fyn kinase substrate
Leverandør: ENZO LIFE SCIENCES
BMLP2150001
ENZOBMLP2150001EA 1650 NOK
ENZOBMLP2150001
Fyn kinase substrate
Proteiner
This peptide, EFGTYGTLSKKK, is derived from a peptide (pF4) discovered, through phage display, to be a very good substrate for Fyn kinase.
  • Its Km is estimated to be 70 µM
The addition of three lysines on the C-terminus allows this peptide to be used in phosphocellulose kinase assays.