PDKtide
Leverandør: ENZO LIFE SCIENCES
BMLP2500100
ENZOBMLP2500100EA 2475 NOK
ENZOBMLP2500100
PDKtide
Proteiner
Protein kinase substrate
This peptide, KTFCGTPEYLAPEVRREPRILSEEEQEMFRDFDYIADWC, is derived from AKT1 and PKN2/PRK2. It is a substrate for PDK1, CDK9/Cyclin T1 and JAK2.
Bestill nå
SPESIFIKASJONER
Protein/peptide name PDKtide
Environmentally Preferable
Arter
Kilde
Renhet 95%