CDK5 substrate,Biotin
Leverandør: ENZO LIFE SCIENCES
BMLP2640001
ENZOBMLP2640001EA 1550 NOK
ENZOBMLP2640001
CDK5 substrate,Biotin
Proteiner
This peptide, Biotin-Ahx-PKTPKKAKKL, is derived from histone H1 and is a substrate for CDK5.
The biotin allows the peptide to be used in kinase assays with streptavidine-bound membranes.