AMARA substrate peptide

Leverandør: ENZO LIFE SCIENCES

BMLP2700001
ENZOBMLP2700001EA 1600 NOK
ENZOBMLP2700001
AMARA substrate peptide
Proteiner

AMPK and SIK kinase substrate


This peptide, AMARAASAAALARRR, is a substrate for AMPK and SIK kinases.

Bestill nå

SPESIFIKASJONER

Protein/peptide name AMARA Substrate Peptide
Environmentally Preferable
Arter
Kilde
Renhet >95% (HPLC)
Formulering Lyophilized

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR