OMNIMMP® RED Fluorogenic Substrate

Leverandør: ENZO LIFE SCIENCES

BMLP2770100
ENZOBMLP2770100EA 2775 NOK
ENZOBMLP2770100
OMNIMMP® RED Fluorogenic Substrate
Proteiner

Highly-quenched, ultra-bright fluorogenic substrate for most MMPs. 6'-TAMRA fluorescence is thoroughly quenched by the TQ3 group until cleavage by MMPs separates the two moieties. The OMNIMMP® RED substrate offers key advantages over other MMP substrates.

Bestill nå

SPESIFIKASJONER

Protein/peptide name RED fluorogenic substrate
Environmentally Preferable
Arter
Kilde
Sekvens TQ3-GABA-Pro-Cha-Abu-Smc-His-Ala-Dab(6'-TAMRA)-Ala-Lys-NH₂
Renhet >90% (HPLC)
Formulering Lyophilised
Molekylvekt 1910.7
Lagringsforhold −20 °C
Oppløslighet Soluble in DMSO (50 mM)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR