Quazinone ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: Ro 13-6438, (R)-6-Chloro-1,5-dihydro-3-methyl- imidazo[2,1-b]quinazolin-2-one

BMLPD1700010
ENZOBMLPD1700010EA 1350 NOK
ENZOBMLPD1700010
Quazinone ≥98% (by HPLC)
Quazinone

Selective inhibitor of cGMP-inhibited phosphodiesterase (PDE III, IC50=0.6µM).


  • PDE inhibitor


White to off-white solid.


Soluble in DMSO (3mg/ml) or 100% ethanol (1mg/ml).

Formel: C₁₁H₁₀ClN₃O
MW: 235,7 g/mol
Lagringstemperatur: Romtemperatur
CAS-nummer: 70018-51-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=12a93a5a077e50671bc1493fc6afc882d07924b8&vid=5d4cc486f6b500ce9aaa4b35e6b7bad017f86bb2&prid=web&clid=SAPDAM

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR