3,4-Dichloroisocoumarin ≥98% (by elemental analysis)

Leverandør: ENZO LIFE SCIENCES

BMLPI1100010
ENZOBMLPI1100010EA 3600 NOK
ENZOBMLPI1100010
3,4-Dichloroisocoumarin ≥98% (by elemental analysis)
3,4-Dichloroisocoumarin

A potent irreversible mechanism based serine protease inhibitor. Inhibits granzymes A, B, and H, cathepsin G, neutrophil elastase and proteinase 3. Inhibits activation of neutral sphingomyelinase and apoptosis triggered by daunorubicin as well as camptothecin-induced apoptosis in HL-60 cells. Inhibits migration of eosinophils through basement membrane components in vitro.


  • Protease inhibitor


White lyophilized solid.

Formel: C₉H₄Cl₂O₂
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036960
CAS-nummer: 51050-59-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb4284f85c172b7edd50ea9f11cd808a08173588&vid=a9f557ab21101dfb6de7226c98a05a5397d346a4&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR