CTT
Leverandør: ENZO LIFE SCIENCES
BMLPI1360001
ENZOBMLPI1360001EA 1900 NOK
ENZOBMLPI1360001
CTT
Proteiner
MMP inhibitor
CTTHWGFTLC is a specific inhibitor of the gelatinases, MMP-2 and MMP-9, with an IC50 of 10-50 µM. Can be used in in vitro studies, zymography and in situ zymography, in tissue culture, and in vivo. Also available, negative control peptide STTHWGFTLS.
Bestill nå
SPESIFIKASJONER
Protein/peptide name CTT
Environmentally Preferable
Arter
Kilde
Formulering Trifluoroacetate salt