Argatroban ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

BMLPI1460010
ENZOBMLPI1460010EA 1625 NOK
ENZOBMLPI1460010
Argatroban ≥98% (by HPLC)
Argatroban

Argatroban is a synthetic peptidomimetic thrombin inhibitor which is in clinical use. It presents significant advantages as an anticoagulant in deep vein thrombosis, stroke prevention in atrial fibrillation and thromboembolic disease and especially in patients with coronary artery disease and heparin-induced thrombocytopenia.


  • Thrombin inhibitor


White to off-white powder.


Soluble in DMSO (25mg/ml) or 100% ethanol (25mg/ml).

Formel: C₂₃H₃₆N₆O₅S
Lagringstemperatur: Fryseskap
MDL nummer: MFCD23102419
CAS-nummer: 74863-84-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR