RWJ-60475 (AM)3
Leverandør: ENZO LIFE SCIENCES
BMLPR1100001
ENZOBMLPR1100001EA 2475 NOK
ENZOBMLPR1100001
RWJ-60475 (AM)3
RWJ-60475 (AM)3
CD45 inhibitor
RWJ-60475-(AM)3 is a cell-permeable acetoxymethyl (AM) ester of RWJ-60475. The AM esters confer cell membrane permeability and are hydrolyzed by intracellular esterases, trapping the active inhibitor within the cell. The IC50 for the hydrolyzed, active inhibitor is 2 µM.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b96a9bdef07750a18bd0c80c608ae8aa373cf42&vid=188dfcdf22203cfc186db2e1ff483150f44d4cdb&prid=web&clid=SAPDAM