Anti-POMP Rabbit Polyclonal Antibody
Leverandør: ENZO LIFE SCIENCES
BMLPW97150025 BMLPW97150100
ENZOBMLPW97150025EA 2800 NOK
ENZOBMLPW97150025 ENZOBMLPW97150100
Anti-POMP Rabbit Polyclonal Antibody
Antistoffer
The assembly of individual mammalian proteasome subunits into catalytically active 20S proteasome is a complex process, with biogenesis of the mammalian 20S proteasome occuring via precursor complexes containing α and unprocessed β subunits. The pro-peptides of the β subunits are thought to prevent premature conversion of the precursor complexes into matured particles and are needed for efficient β subunit incorporation. The complex biogenesis is tightly regulated, requiring additional components such as the maturation factor POMP (proteasome maturation protein), an ubiquitous protein inMore Product Information