Human Recombinant 14-3-3sigma (from E. coli), HIS Tag

Leverandør: ENZO LIFE SCIENCES

BMLSE4880100
ENZOBMLSE4880100EA 4475 NOK
ENZOBMLSE4880100
Human Recombinant 14-3-3sigma (from E. coli), HIS Tag
Proteiner

Produced in E. coli. Human 14-3-3s is fused at the C-terminus to a His-tag.

Bestill nå

SPESIFIKASJONER

Protein/peptide name 14-3 -3 sigma
Environmentally Preferable
Protein/peptide type Recombinant
Arter Human
Kilde E. coli
Renhet >90% (SDS-PAGE)
Tag sekvens HIS Tag

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR