Human recombinant PKCtheta
Leverandør: ENZO LIFE SCIENCES
BMLSE5170005
ENZOBMLSE5170005EA 4100 NOK
ENZOBMLSE5170005
Human recombinant PKCtheta
Proteiner
Produced in insect cells. Full length active enzyme fused at the N-terminus to a GST-tag.