Human recombinant DPP8
Leverandør: ENZO LIFE SCIENCES
BMLSE5270010
ENZOBMLSE5270010EA 8100 NOK
ENZOBMLSE5270010
Human recombinant DPP8
Proteiner
Produced in insect cells. Active recombinant human DPP8 with an N-terminal purification tag.