Human recombinant neprilysin (from insect cells (Sf9)), HIS tag
Leverandør: ENZO LIFE SCIENCES
BMLSE5320010
ENZOBMLSE5320010EA 6125 NOK
ENZOBMLSE5320010
Human recombinant neprilysin (from insect cells (Sf9)), HIS tag
Proteiner
Highly active
Produced in Sf9 cells.
Neprilysin is a glycosylated zinc endopeptidase whose physiological targets include bradykinin, glucagon, GLP-1, and neuropeptides. Preferential cleavage of polypeptides between hydrophobic residues, particularly with Phe or Tyr at P1.