Recombinant ubiquitin (from E. coli), rhodamine

Leverandør: ENZO LIFE SCIENCES

BMLSE7610025
ENZOBMLSE7610025EA 3050 NOK
ENZOBMLSE7610025
Recombinant ubiquitin (from E. coli), rhodamine
Proteiner

Ubiquitin-rhodamine is a high purity industry standard substrate for studying deubiquitinylating activity where detection sensitivity or continuous monitoring of activity.


  • Molecular weight: ~8,9 kDa


It is prepared by the C-terminal derivatisation of ubiquitin with modified rhodamine-110 and has been shown to be a useful and sensitive fluorogenic substrate for wide range of deubiquitinylating enzymes (DUB's), including ubiquitin C-terminal hydrolases (UCH's) and ubiquitin specific proteases (USP's)

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR