Sphingosine-1-phosphate ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES
Warning

Synonymer: S1P

BMLSL1400001
ENZOBMLSL1400001EA 2350 NOK
ENZOBMLSL1400001
Sphingosine-1-phosphate ≥98% (by TLC)
Sphingosine-1-phosphate

Endogenous signaling molecule.


Sphingosine-1-phosphate (1-SPP) is a putative second messenger that mobilizes calcium from intracellular stores via an IP3 independent pathway, activation of phospholipase D and stimulation of quiescent Swiss 3T3 fibroblast growth. Regulates a variety of cellular responses, including survival, cytoskeletal remodeling, chemotaxis etc. via the activation of cell surface EDG receptors. 1-SPP also promotes angiogenesis.


Sparingly soluble in 1:1 ethanol:water. Addition of small amounts of acetic acid may help dissolve this product. Soluble in warm methanol.

Formel: C₁₈H₃₈NO₅P
MW: 379.478 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00270077
CAS-nummer: 26993-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e51030518076766544aa75137a50a46d9dd507bb&vid=6fd056bcd7a28ca8b0ff509dafe0053e9f65e0e1&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR