α-Galactosylceramide (synthetic) ≥99% (by HPLC)

Leverandør: ENZO LIFE SCIENCES

Synonymer: alpha-GalCer, alpha-Galactosylceramide, N-((2S,3S,4R)-3,4-Dihydroxy-1-(((2S,3R,4S,5R,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)octadecan-2-yl)hexacosanamide, KRN7000, a-​Galactosylceramide, (2S,3S,4R)-1-O-(α-D-Galactosyl)-N-hexacosanoyl-2-amino-1,3,4-octadecanetriol

BMLSL2320100 BMLSL2321000
ENZOBMLSL2320100EA 4375 NOK
ENZOBMLSL2320100 ENZOBMLSL2321000
α-Galactosylceramide (synthetic) ≥99% (by HPLC)
α-Galactosylceramide

MHC ligand.


KRN7000 is a synthetic analog of α-galactosylceramide and the marine natural product agelasphin. It is a specific ligand for human and mouse NKT cells and remains the best studied ligand of the lipid-binding MHC class I-like protein CD1d. It protects against LPS-induced shock and displays potent antitumor activity in various in vivo models.


KRN7000 is inherently an extremely hydrophobic molecule, therefore, almost all the methods for solubilizing this material in primarily aqueous media will contain at least some detergent. For cell culture or other in vivo models, it is recommended that the KRN7000 be initially dissolved in a 2:1 mixture of chloroform and methanol.

Formel: C₅₀H₉₉NO₉
MW: 858.34 g/mol
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00939559
CAS-nummer: 158021-47-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7334f05253383bf78b11488c03d8c2b4ce01235a&vid=d7505aa4e2fb9b4fc828461371cce30ab4b9f384&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥99% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR