Bryostatin 1 ≥98% (by HPLC)

Leverandør: ENZO LIFE SCIENCES
Danger

BMLST1030010
ENZOBMLST1030010EA 4775 NOK
ENZOBMLST1030010
Bryostatin 1 ≥98% (by HPLC)
Bryostatin 1

PKC activator.


A structurally unique marine natural product that binds to and activates PKC at picomolar concentrations, but is not a carcinogen or a complete tumor promoter. In many systems it induces only a subset of the responses to PMA and blocks those which it does not induce. It is markedly more potent than PMA for translocating PKCδ and -ε.


Soluble in DMSO (25 mg/ml) or 100% ethanol (25 mg/ml).

Formel: C₄₇H₆₈O₁₇
MW: 905.044 g/mol
Lagringstemperatur: Fryseskap
CAS-nummer: 83314-01-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=26eb281ee72541a5743fe008f0415698814c27de&vid=e79faec61154296560075f18b651bbfab73492ee&prid=web&clid=SAPDAM

Bestill nå

Spesifikasjon Testresultater

Purity ≥98% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR