PI 3K SH2 OMT
Leverandør: ENZO LIFE SCIENCES
BMLST4250500
ENZOBMLST4250500EA 2475 NOK
ENZOBMLST4250500
PI 3K SH2 OMT
Proteiner
PI3 kinase inhibitor
This unique SH2 domain ligand is derived from the sequence flanking Tyr751 of the PDGF receptor. It inhibits the PI 3-kinase p85 C-terminal SH2 domain (IC50=14,2 µM). It contains the nonhydrolyzable phosphotyrosine mimetic L-O-(2-malonyl)tyrosine (OMT) and is provided as the trimethyl ester for greater cell permeability.
Bestill nå
SPESIFIKASJONER
Protein/peptide name PI 3K SH2 OMT
Environmentally Preferable
Arter
Kilde