Bovine serum albumin (BSA) (nitrated) (from serum)
Leverandør: ENZO LIFE SCIENCES
BMLSW1250100
ENZOBMLSW1250100EA 1950 NOK
ENZOBMLSW1250100
Bovine serum albumin (BSA) (nitrated) (from serum)
Proteiner
Nitrated bovine serum albumin for use as a positive control for antibodies to nitrotyrosine.