Latrunculin A ≥98% (by TLC)

Leverandør: ENZO LIFE SCIENCES

BMLT1190100 BMLT1190500
ENZOBMLT1190100EA 2025 NOK
ENZOBMLT1190100 ENZOBMLT1190500
Latrunculin A ≥98% (by TLC)
Latrunculin A

Ultra-pure; inhibits actin polymerization


Inhibits actin polymerization, microfilament organzation and microfilament-mediated processes. Acts via a different mechanism and 10- to 20-fold more potently than cytochalasins. Inhibits phagocytosis by macrophages. Exposure to 0.2 mg/ml for one hour produces maximal change in cell morphology.

Formel: C₂₂H₃₁NO₅S
CAS-nummer: 76343-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ecf1ee0ecf889b337739abc75420a9544058d88&vid=da41150ef3e1a659af05094d3a91f581919c8143&prid=web&clid=SAPDAM

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR