Ubiquitin activating kit

Leverandør: ENZO LIFE SCIENCES

BMLUW0400A0001
ENZOBMLUW0400A0001EA 2250 NOK
ENZOBMLUW0400A0001
Ubiquitin activating kit
Assay Cellulær analyse

Ubiquitin Activating Kit for activation of ubiquitin for use in ubiquitinylation experiments. The kit provides the means of generating thioester linked, activated ubiquitin-E1 conjugates, utilising the first step in the ubiquitin cascade, for investigation of ubiquitin activation, subsequent ubiquitin transfer to/interaction with E2 conjugating enzymes, and their use in the ubiquitinylation of E3 ligases and target substrate proteins. Ubiquitin antibody provided allows high sensitivity detection of ubiquitin and ubiquitin-conjugates.


  • For use with wild type or mutant E2 enzymes in E1 initiated/mediated reactions
  • Contains highly sensitive ubiquitin antibody for detection of ubiquitin and ubiquitin conjugates


Uses:
Activation of Ubiquitin for conjugation to wild type or mutant E2 enzymes (user supplied) via thioester bond formation.
Use of Ub-E2 conjugates produced in the subsequent ubiquitin modification of specific target proteins in presence of a dedicated ubiquitin E3 ligase.
Investigation of ubiquitin activation by E1 activating enzyme.
Substitution of the wild type ubiquitin provided with ubiquitin mutants or derivatives (e.g. biotinylated-ubiquitin, Prod. No. BML-UW8705) allowing their activation and subsequent utility in the ubiquitin cascade.


Bestillingsinformasjon: Contains 20X Ubiquitin Activating Enzyme Solution, Ubiquitin Solution, Ubiquitin Antibody Solution, Ubiquitin protein conjugates rabbit PAb, 20X Mg-ATP Solution, 2X Non-reducing Gel Loading Buffer, 10X Ubiquitinylation Buffer.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR