Tetra-ubiquitin (linear) (from Escherichia coli)
Leverandør: ENZO LIFE SCIENCES
BMLUW07850100
ENZOBMLUW07850100EA 1650 NOK
ENZOBMLUW07850100
Tetra-ubiquitin (linear) (from Escherichia coli)
Proteiner
Linear polyubiquitin chains are useful tools for investigating, amongst other things, the specificity and reactivity of deubiquitinylating enzymes (DUBs) and the recognition and interaction of linear polyubiquitin modified proteins with by ubiquitin binding domain (UBDs) containing proteins.