Anti-NGAL Mouse Monoclonal Antibody [clone: 68]

Leverandør: ENZO LIFE SCIENCES
BPDABS063681
ENZOBPDABS063681EA 49975 NOK
ENZOBPDABS063681
Anti-NGAL Mouse Monoclonal Antibody [clone: 68]
Antistoffer
Recommended Applications: ELISA, IHC

Species reactivity: Rhesus monkey, Cynomolgus

Type: Primary
Antigen: NGAL
Clonality: Monoclonal
Clone: 68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1κ
Reactivity: Rhesus monkey, Cynomolgus
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR