Human IgE (Non-Immune, Azide)
Leverandør: ENZO LIFE SCIENCES
BPDDIAHE1A04
ENZOBPDDIAHE1A04EA 26925 NOK
ENZOBPDDIAHE1A04
Human IgE (Non-Immune, Azide)
Proteiner
Purified human IgE with kappa light chains produced in vitro from a monoclonal hybridoma.
Bestill nå
SPESIFIKASJONER
Protein/peptide name IgE (non-immune, azide)
Environmentally Preferable
Protein synonyms Immunoglobulin E
Arter Human
Kilde
Konjugering Unconjugated
Formulering Liquid. In 0,01 M phosphate buffer, pH 7,4, containing 0,15 M sodium chloride and 0,09% sodium azide.
Frakttemp. 20