Wash buffer salts, lyophilised, Enzo Life Sciences
Leverandør: ENZO LIFE SCIENCES
ENZ33802
ENZOENZ33802EA 930 NOK
ENZOENZ33802
Wash buffer salts, lyophilised, Enzo Life Sciences
Buffere Buffere, cellekultur
Wash Buffer Salts can be used in tissue preparations, immunohistochemical staining procedures and in assays that require HRP-linked detection reagents.
  • Each packet of salts yields 1 liter of buffered saline
  • pH should be 7.1 ± 0.1
Bestill nå
SPESIFIKASJONER
Størrelse 3 L
Environmentally Preferable
Formulering Lyophilised
Sensitivitet Dissolve entire contents of packet in 1 L of distilled or deionised water. The pH of the solution should be 7,1 ±0,1.