In situ hybridisation wash reagent
Leverandør: ENZO LIFE SCIENCES
ENZ33809
ENZOENZ33809EA 1950 NOK
ENZOENZ33809
In situ hybridisation wash reagent
Buffere Buffere, elektroforese Buffere, hybridisering
Hybridisation wash reagent provides a high quality hybridisation wash solution for in situ hybridisation analyses using DNA probes labeled with biotin, digoxigenin or fluorescein.
Formulation: Liquid, PBS containing deionised formamide.