Cyanine 3-UTP (enhanced)
Leverandør: ENZO LIFE SCIENCES
ENZ42505
ENZOENZ42505EA 5475 NOK
ENZOENZ42505
Cyanine 3-UTP (enhanced)
Nukleinsyre reagenser Nukleotider
Cyanine 3-UTP (Enhanced) can replace UTP as a substrate for T7 RNA polymerase in labeling systems that generate labeled probes through in vitro transcription. Probes generated by these methods are suitable for multicolor fluorescence analysis, specifically dual-color expression arrays in conjunction with cyanine 3-UTP .
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Applikasjon
Konsentrasjon 10 mM
Formulering Liquid. Solution in water.
Renhet ≥93% (HPLC)