Bio-11-CTP

Leverandør: ENZO LIFE SCIENCES

ENZ42818
ENZOENZ42818EA 4300 NOK
ENZOENZ42818
Bio-11-CTP
Nukleinsyre reagenser Nukleotider

Bio-11-CTP (Biotin-11-cytidine-5’-triphosphate) can replace CTP for in vitro transcription reaction catalysed by T3, T7 or SP6 RNA polymerases.


  • Appearance: Clear, colourless liquid
  • Purity: ≥93% (HPLC)
  • Concentration: 10 mM
  • Extinction coefficient: 10700 M⁻¹ cm⁻¹ (240 nm, pH 7,0)


The biotin-labelled RNA transcripts produced by these reactions are suitable for a wide range of applications such as nucleic acid hybridisation, sequencing, and genome analysis.


The biotin-labelled hybridised probes can be detected by a reporter molecule linked to streptavidin, avidin, or anti-biotin antibody. Such a complex can be detected directly, e.g. by excitation of a fluorophore conjugated to streptavidin, or indirectly, e.g. using an enzyme conjugate that can produce an insoluble coloured precipitate.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46efb2e297f1135678942a325eb8e055020288e&vid=a9ac475218ed4b4f916d55ae2f3dbd2d60e5df73&prid=web&clid=SAPDAM

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR